siRNA Details
Gene Symbol: LOC680200 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001139491.1 | NP_001132963.1 | 4 | 1234 | ||
GenBank mRNA | BC167062.1 | - | 4 | 1234 | 9024521 |
Gene Symbol: Rsl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006232102.2 | XP_006232164.1 | 4 | 1588 | ||
XM_006232103.2 | XP_006232165.1 | 4 | 1857 | |||
XM_006232104.2 | XP_006232166.1 | 4 | 1312 | |||
XM_006232105.2 | XP_006232167.1 | 4 | 1612 | |||
XM_006232106.2 | XP_006232168.1 | 4 | 1320 | |||
XM_008760847.1 | XP_008759069.1 | 4 | 1900 |
Gene Symbol: Zfp455 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173314.2 | NP_775436.2 | 4 | 1282 | ||
XM_006232101.2 | XP_006232163.1 | 4 | 1518 | |||
GenBank mRNA | BC161833.1 | - | 5 | 1114 | ||
U67083.1 | - | 1270 |
