siRNA Details
Gene Symbol: LOC286992 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173326.1 | NP_775448.1 | 556 | |||
GenBank mRNA | AF012282.1 | - | 556 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC286992 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173326.1 | NP_775448.1 | 556 | |||
GenBank mRNA | AF012282.1 | - | 556 |