siRNA Details
| Gene Symbol: C6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_176074.3 | NP_788263.1 | 5, 6 | 736 | ||
| XM_008760747.1 | XP_008758969.1 | 6, 7 | 921 | |||
| XM_008760749.1 | XP_008758971.1 | 6, 7 | 906 | |||
| XM_008760750.1 | XP_008758972.1 | 6, 7 | 924 | |||
| GenBank mRNA | AY230250.1 | - | 5, 6 | 738 | ||

C6
Genomic Map