siRNA Details
| Gene Symbol: HOXD8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001199746.1 | NP_001186675.1 | 1 | 710 | ||
| NM_019558.3 | NP_062458.1 | 1 | 710 | |||
| GenBank mRNA | AL530925.3 | - | 1 | 710 | ||
| BC038709.1 | - | 1 | 560 | 4614350 | ||
| BC090853.1 | - | 2 | 409 | 6180525 | ||

HOXD8
Genomic Map