siRNA Details
| Gene Symbol: Sept9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001113497.1 | NP_001106969.1 | 220 | |||
| NM_031837.2 | NP_114025.2 | 1108 | ||||
| NM_176856.2 | NP_789826.2 | 1227 | ||||
| GenBank mRNA | AF170253.1 | - | 1228 | |||
| AF173899.1 | - | 1107 | ||||
| AF180525.1 | - | 2 | 387 | |||
| AF180526.1 | - | 208 | ||||
| BC166602.1 | - | 773 | ||||
| CF977797.1 | - | 2 | 246 | |||

Sept9
Genomic Map