siRNA Details
Gene Symbol: Tp53inp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181084.2 | NP_851598.1 | 2 | 271 | ||
GenBank mRNA | AB107917.1 | - | 2 | 271 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Tp53inp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181084.2 | NP_851598.1 | 2 | 271 | ||
GenBank mRNA | AB107917.1 | - | 2 | 271 |