siRNA Details
| Gene Symbol: Nudt6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181363.2 | NP_852028.1 | 1 | 174 | ||
| GenBank mRNA | AF188995.1 | - | 1 | 153 | ||
| AF188996.1 | - | 1 | 153 | |||
| U58289.1 | - | 1 | 174 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nudt6| Gene Symbol: Nudt6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181363.2 | NP_852028.1 | 1 | 174 | ||
| GenBank mRNA | AF188995.1 | - | 1 | 153 | ||
| AF188996.1 | - | 1 | 153 | |||
| U58289.1 | - | 1 | 174 | |||