siRNA Details
| Gene Symbol: Lgsn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181383.2 | NP_852048.1 | 2 | 108 | ||
| GenBank mRNA | AY280501.1 | - | 2 | 108 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lgsn| Gene Symbol: Lgsn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181383.2 | NP_852048.1 | 2 | 108 | ||
| GenBank mRNA | AY280501.1 | - | 2 | 108 | ||