siRNA Details
| Gene Symbol: Tmem184c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178330.1 | NP_847900.1 | 2, 3 | 718 | ||
| XM_006255422.2 | XP_006255484.1 | 3, 4 | 430 | |||
| XM_008772475.1 | XP_008770697.1 | 3, 4 | 1329 | |||
| GenBank mRNA | AY228474.1 | - | 2, 3 | 718 | ||
| BC089112.1 | - | 2, 3 | 728 | 7316181 | ||

Tmem184c
Genomic Map