siRNA Details
| Gene Symbol: Egln1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008772679.1 | XP_008770901.1 | 4, 5 | 1631 | ||
| GenBank mRNA | AY228140.1 | - | 2, 3 | 416 | ||
| BC081694.1 | - | 2, 3 | 660 | |||
| Gene Symbol: LOC690182 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008759848.1 | XP_008758070.1 | 1 | 656 | ||

Egln1
Genomic Map