siRNA Details
| Gene Symbol: Klhl24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181473.2 | NP_852138.2 | 2 | 289 | ||
| GenBank mRNA | AB101615.1 | - | 1 | 141 | ||
| BC092204.1 | - | 2 | 289 | 7312023 | ||
| FQ219171.1 | - | 3 | 372 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Klhl24| Gene Symbol: Klhl24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181473.2 | NP_852138.2 | 2 | 289 | ||
| GenBank mRNA | AB101615.1 | - | 1 | 141 | ||
| BC092204.1 | - | 2 | 289 | 7312023 | ||
| FQ219171.1 | - | 3 | 372 | |||