siRNA Details
Gene Symbol: Cd2ap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181475.2 | NP_852140.1 | 2 | 204 | ||
GenBank mRNA | AY114147.1 | - | 2 | 200 | ||
AY205155.1 | - | 2 | 204 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Cd2ap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181475.2 | NP_852140.1 | 2 | 204 | ||
GenBank mRNA | AY114147.1 | - | 2 | 200 | ||
AY205155.1 | - | 2 | 204 |