siRNA Details
| Gene Symbol: Slc38a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181090.2 | NP_851604.1 | 5, 6 | 790 | ||
| XM_006242282.1 | XP_006242344.1 | 5, 6 | 830 | |||
| XM_006242283.1 | XP_006242345.1 | 4, 5 | 714 | |||
| GenBank mRNA | AF173682.2 | - | 5, 6 | 591 | ||
| AF249673.1 | - | 5, 6 | 790 | |||
| AF273024.1 | - | 5, 6 | 625 | |||

Slc38a2
Genomic Map