siRNA Details
| Gene Symbol: Rtn4rl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181377.3 | NP_852042.1 | 2 | 981 | ||
| GenBank mRNA | AF532861.1 | - | 2 | 736 | ||
| AY311478.1 | - | 3 | 844 | |||
| BC129084.1 | - | 2 | 981 | 8363864 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rtn4rl1| Gene Symbol: Rtn4rl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181377.3 | NP_852042.1 | 2 | 981 | ||
| GenBank mRNA | AF532861.1 | - | 2 | 736 | ||
| AY311478.1 | - | 3 | 844 | |||
| BC129084.1 | - | 2 | 981 | 8363864 | ||