siRNA Details
| Gene Symbol: Cox8b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012786.3 | NP_036918.2 | 1 | 132 | ||
| GenBank mRNA | FM054348.1 | - | 1 | 132 | ||
| X64827.1 | - | 1 | 127 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cox8b| Gene Symbol: Cox8b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012786.3 | NP_036918.2 | 1 | 132 | ||
| GenBank mRNA | FM054348.1 | - | 1 | 132 | ||
| X64827.1 | - | 1 | 127 | |||