siRNA Details
Gene Symbol: Oprm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001038597.2 | NP_001033686.1 | 1, 2 | 499 | ||
NM_001038599.2 | NP_001033688.2 | 1, 2 | 499 | |||
NM_001038600.2 | NP_001033689.1 | 1, 2 | 499 | |||
NM_001038601.2 | NP_001033690.1 | 1, 2 | 499 | |||
NM_013071.2 | NP_037203.1 | 1, 2 | 499 | |||
XM_008758731.1 | XP_008756953.1 | 1, 2 | 645 | |||
GenBank mRNA | AY225402.1 | - | 1, 2 | 330 | ||
AY225403.1 | - | 1, 2 | 330 | |||
AY309000.1 | - | 1, 2 | 330 | |||
AY309002.1 | - | 1, 2 | 324 | |||
AY309003.1 | - | 1, 2 | 330 | |||
AY309004.1 | - | 1, 2 | 330 | |||
D16349.1 | - | 1, 2 | 475 | |||
EU024651.1 | - | 511 | ||||
EU024652.1 | - | 700 | ||||
EU340244.1 | - | 519 | ||||
EU340245.1 | - | 775 | ||||
EU340246.1 | - | 961 | ||||
L13069.1 | - | 1, 2 | 501 | |||
L20684.1 | - | 1, 2 | 322 | |||
L22455.1 | - | 1, 2 | 461 | |||
U02083.1 | - | 1, 2 | 502 | |||
U35424.1 | - | 1, 2 | 361 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
