siRNA Details
| Gene Symbol: Otx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013109.2 | NP_037241.1 | 2 | 226 | ||
| GenBank mRNA | FQ212299.1 | - | 430 | |||
| L32602.1 | - | 2 | 227 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Otx1| Gene Symbol: Otx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013109.2 | NP_037241.1 | 2 | 226 | ||
| GenBank mRNA | FQ212299.1 | - | 430 | |||
| L32602.1 | - | 2 | 227 | |||