siRNA Details
| Gene Symbol: TSHZ3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AB040907.1 | - | 2 | 868 | ||
| AK291466.1 | - | 2 | 890 | |||
| AL136805.1 | - | 1 | 349 | |||
| BC127095.1 | - | 1 | 349 | 40123924 | ||
| BC127096.1 | - | 1 | 349 | 40123925 | ||
| BX648745.1 | - | 2 | 891 | |||
| CN256026.1 | - | 1 | 659 | |||

TSHZ3
Genomic Map