siRNA Details
| Gene Symbol: Slc7a5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017353.1 | NP_059049.1 | 1 | 215 | ||
| GenBank mRNA | AB015432.1 | - | 1 | 215 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc7a5| Gene Symbol: Slc7a5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017353.1 | NP_059049.1 | 1 | 215 | ||
| GenBank mRNA | AB015432.1 | - | 1 | 215 | ||