siRNA Details
| Gene Symbol: Pla2g2c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019202.2 | NP_062075.1 | 4 | 490 | ||
| XM_006239142.2 | XP_006239204.1 | 4 | 508 | |||
| XM_006239143.2 | XP_006239205.1 | 3 | 269 | |||
| XM_008764212.1 | XP_008762434.1 | 4 | 583 | |||
| XM_008764213.1 | XP_008762435.1 | 4 | 361 | |||
| XM_008764214.1 | XP_008762436.1 | 4 | 2032 | |||
| GenBank mRNA | BC097325.1 | - | 4 | 490 | 7456563 | |
| U07798.1 | - | 4 | 817 | |||

Pla2g2c
Genomic Map