siRNA Details
| Gene Symbol: BCL7A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024808.2 | NP_001019979.1 | 2, 3 | 379 | ||
| NM_020993.4 | NP_066273.1 | 2, 3 | 379 | |||
| GenBank mRNA | AK296159.1 | - | 2, 3 | 381 | ||
| AK316486.1 | - | 2, 3 | 277 | |||
| BC094723.1 | - | 2, 3 | 190 | 5300018 | ||
| BC113627.1 | - | 2, 3 | 203 | 8322679 | ||
| BM801030.1 | - | 338 | ||||
| X89984.1 | - | 2, 3 | 1126 | |||

BCL7A
Genomic Map