siRNA Details
| Gene Symbol: Fxyd7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022008.1 | NP_071291.1 | 3, 4 | 232 | ||
| XM_006228843.2 | XP_006228905.1 | 3, 4 | 312 | |||
| GenBank mRNA | AW914994.1 | - | 3, 4 | 232 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fxyd7| Gene Symbol: Fxyd7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022008.1 | NP_071291.1 | 3, 4 | 232 | ||
| XM_006228843.2 | XP_006228905.1 | 3, 4 | 312 | |||
| GenBank mRNA | AW914994.1 | - | 3, 4 | 232 | ||