siRNA Details
| Gene Symbol: Mgp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012862.1 | NP_036994.1 | 3, 4 | 190 | ||
| GenBank mRNA | BC086394.1 | - | 3, 4 | 222 | 7303359 | |
| FQ221152.1 | - | 1, 2 | 28 | |||
| FQ221407.1 | - | 1, 2 | 28 | |||
| FQ224403.1 | - | 3, 4 | 247 | |||
| FQ229124.1 | - | 1, 2 | 28 | |||
| J03026.1 | - | 3, 4 | 190 | |||

Mgp
Genomic Map