siRNA Details
| Gene Symbol: Slc30a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001083122.1 | NP_001076591.1 | 2 | 237 | ||
| NM_012890.2 | NP_037022.1 | 1 | 176 | |||
| GenBank mRNA | BC061997.1 | - | 1 | 179 | ||
| BC168237.1 | - | 2 | 230 | 7938890 | ||
| CB778290.1 | - | 2 | 240 | |||
| U50927.1 | - | 1 | 176 | |||

Slc30a2
Genomic Map