siRNA Details
| Gene Symbol: Pemt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013003.1 | NP_037135.1 | 5 | 481 | ||
| GenBank mRNA | BC091162.1 | - | 5 | 521 | 7325819 | |
| L14441.1 | - | 5 | 481 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pemt| Gene Symbol: Pemt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013003.1 | NP_037135.1 | 5 | 481 | ||
| GenBank mRNA | BC091162.1 | - | 5 | 521 | 7325819 | |
| L14441.1 | - | 5 | 481 | |||