siRNA Details
| Gene Symbol: Btg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017259.1 | NP_058955.1 | 2 | 241 | ||
| GenBank mRNA | BC072493.1 | - | 2 | 200 | 7097327 | |
| M60921.1 | - | 2 | 241 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Btg2| Gene Symbol: Btg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017259.1 | NP_058955.1 | 2 | 241 | ||
| GenBank mRNA | BC072493.1 | - | 2 | 200 | 7097327 | |
| M60921.1 | - | 2 | 241 | |||