siRNA Details
Gene Symbol: Atp5g1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017311.1 | NP_059007.1 | 4 | 226 | ||
XM_006247188.2 | XP_006247250.1 | 4 | 269 | |||
XM_006247189.1 | XP_006247251.1 | 4 | 374 | |||
GenBank mRNA | D13123.1 | - | 4 | 226 | ||
FQ209790.1 | - | 4 | 244 | |||
FQ210883.1 | - | 3 | 190 | |||
FQ216886.1 | - | 4 | 244 | |||
FQ217997.1 | - | 4 | 240 |
