siRNA Details
| Gene Symbol: Gosr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031685.2 | NP_113873.1 | 2 | 97 | ||
| GenBank mRNA | AF007549.1 | - | 2 | 111 | ||
| BC061994.1 | - | 2 | 97 | 5599557 | ||
| U91539.1 | - | 2 | 96 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gosr2| Gene Symbol: Gosr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031685.2 | NP_113873.1 | 2 | 97 | ||
| GenBank mRNA | AF007549.1 | - | 2 | 111 | ||
| BC061994.1 | - | 2 | 97 | 5599557 | ||
| U91539.1 | - | 2 | 96 | |||