siRNA Details
Gene Symbol: Smpx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053395.1 | NP_445847.1 | 4 | 350 | ||
XM_006256945.2 | XP_006257007.1 | 4 | 319 | |||
GenBank mRNA | AF364071.1 | - | 4 | 350 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Smpx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053395.1 | NP_445847.1 | 4 | 350 | ||
XM_006256945.2 | XP_006257007.1 | 4 | 319 | |||
GenBank mRNA | AF364071.1 | - | 4 | 350 |