siRNA Details
| Gene Symbol: Twist1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053530.2 | NP_445982.1 | 1 | 555 | ||
| GenBank mRNA | AF266260.1 | - | 1 | 50 | ||
| BC166412.1 | - | 1 | 685 | 9033082 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Twist1| Gene Symbol: Twist1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053530.2 | NP_445982.1 | 1 | 555 | ||
| GenBank mRNA | AF266260.1 | - | 1 | 50 | ||
| BC166412.1 | - | 1 | 685 | 9033082 | ||