siRNA Details
| Gene Symbol: Cox17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053540.2 | NP_445992.1 | 2 | 218 | ||
| GenBank mRNA | AB032178.1 | - | 2 | 162 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_001017429, AK160687, BG792513, AK158051, AK146388, and others.

Cox17
Genomic Map