siRNA Details
Gene Symbol: Dmrt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053706.1 | NP_446158.1 | 1 | 413 | ||
XM_008760301.1 | XP_008758523.1 | 1 | 503 | |||
GenBank mRNA | AF379608.1 | - | 1 | 413 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Dmrt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053706.1 | NP_446158.1 | 1 | 413 | ||
XM_008760301.1 | XP_008758523.1 | 1 | 503 | |||
GenBank mRNA | AF379608.1 | - | 1 | 413 |