siRNA Details
| Gene Symbol: Kcnk4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053804.2 | NP_446256.2 | 1 | 129 | ||
| XM_006230634.2 | XP_006230696.1 | 2 | 1427 | |||
| XM_008760054.1 | XP_008758276.1 | 2 | 1170 | |||
| XM_008760055.1 | XP_008758277.1 | 2 | 1041 | |||
| XM_008760056.1 | XP_008758278.1 | 2 | 247 | |||
| GenBank mRNA | AF302842.2 | - | 1 | 129 | ||

Kcnk4
Genomic Map