siRNA Details
| Gene Symbol: F2rl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053808.1 | NP_446260.1 | 2 | 111 | ||
| GenBank mRNA | AF269246.2 | - | 2 | 111 | ||
| AF310216.1 | - | 2 | 138 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

F2rl3| Gene Symbol: F2rl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053808.1 | NP_446260.1 | 2 | 111 | ||
| GenBank mRNA | AF269246.2 | - | 2 | 111 | ||
| AF310216.1 | - | 2 | 138 | |||