siRNA Details
| Gene Symbol: Dynlrb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_131910.3 | NP_571985.1 | 2, 3 | 202 | ||
| GenBank mRNA | AF073839.2 | - | 2, 3 | 125 | ||
| AY026512.1 | - | 2, 3 | 202 | |||
| BC058437.1 | - | 2, 3 | 143 | 6921076 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_025947, AK166685, AK008214, AK003435, AK002359, and others.

Dynlrb1
Genomic Map