siRNA Details
Gene Symbol: Cxcl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030845.1 | NP_110472.1 | 2 | 203 | ||
GenBank mRNA | D11444.1 | - | 2 | 188 | ||
M86536.1 | - | 2 | 203 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Cxcl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030845.1 | NP_110472.1 | 2 | 203 | ||
GenBank mRNA | D11444.1 | - | 2 | 188 | ||
M86536.1 | - | 2 | 203 |