siRNA Details
| Gene Symbol: Fads2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031344.2 | NP_112634.1 | 1, 2 | 370 | ||
| GenBank mRNA | AB021980.1 | - | 1, 2 | 300 | ||
| BC081776.1 | - | 1, 2 | 370 | 7128630 | ||
| FQ210893.1 | - | 1, 2 | 299 | |||
| HQ909027.1 | - | 1, 2 | 204 | |||

Fads2
Genomic Map