siRNA Details
| Gene Symbol: Fxyd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031648.2 | NP_113836.2 | 6 | 320 | ||
| XM_006228838.2 | XP_006228900.1 | 7 | 540 | |||
| XM_006228839.2 | XP_006228901.1 | 6 | 315 | |||
| XM_008759184.1 | XP_008757406.1 | 6 | 329 | |||
| GenBank mRNA | CB712570.1 | - | 6 | 320 | ||
| FM109156.1 | - | 6 | 274 | |||
| FQ211349.1 | - | 6 | 331 | |||
| FQ224131.1 | - | 6 | 283 | |||
| FQ224338.1 | - | 6 | 366 | |||
| U72246.1 | - | 5 | 234 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_052992, NM_019503, NM_052991, NM_194321, CA459152, and others.

Fxyd1
Genomic Map