siRNA Details
| Gene Symbol: Aqp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031703.1 | NP_113891.1 | 4 | 514 | ||
| GenBank mRNA | BC127490.1 | - | 4 | 496 | 8366493 | |
| D17695.1 | - | 4 | 429 | |||
| L35108.1 | - | 4 | 514 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Aqp3| Gene Symbol: Aqp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031703.1 | NP_113891.1 | 4 | 514 | ||
| GenBank mRNA | BC127490.1 | - | 4 | 496 | 8366493 | |
| D17695.1 | - | 4 | 429 | |||
| L35108.1 | - | 4 | 514 | |||