siRNA Details
Gene Symbol: Pex11a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053487.2 | NP_445939.1 | 2 | 246 | ||
XM_008759560.1 | XP_008757782.1 | 1 | 98 | |||
GenBank mRNA | AJ224120.1 | - | 2 | 278 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Pex11a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053487.2 | NP_445939.1 | 2 | 246 | ||
XM_008759560.1 | XP_008757782.1 | 1 | 98 | |||
GenBank mRNA | AJ224120.1 | - | 2 | 278 |