siRNA Details
| Gene Symbol: Twist1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053530.2 | NP_445982.1 | 1 | 615 | ||
| GenBank mRNA | AF266260.1 | - | 1 | 110 | ||
| BC166412.1 | - | 1 | 745 | 9033082 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_011658, BC083139, BC033434, M63650.

Twist1
Genomic Map