siRNA Details
| Gene Symbol: Fxyd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017349.2 | NP_059045.2 | 2, 3 | 142 | ||
| NM_145717.1 | NP_663769.1 | 3, 4 | 186 | |||
| XM_006242911.2 | XP_006242973.1 | 3, 4 | 343 | |||
| XM_006242912.2 | XP_006242974.1 | 3, 4 | 291 | |||
| GenBank mRNA | AF129400.1 | - | 3, 4 | 186 | ||
| AF233060.1 | - | 2, 3 | 142 | |||
| X70062.1 | - | 3, 4 | 150 | |||

Fxyd2
Genomic Map