siRNA Details
| Gene Symbol: Mrgprf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153722.2 | NP_714944.1 | 3 | 323 | ||
| GenBank mRNA | M35297.1 | - | 326 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mrgprf| Gene Symbol: Mrgprf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153722.2 | NP_714944.1 | 3 | 323 | ||
| GenBank mRNA | M35297.1 | - | 326 | |||
Set Membership: |
|