siRNA Details
Gene Symbol: LOC299271 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181630.2 | NP_853661.1 | 2 | 332 | ||
GenBank mRNA | AY113703.1 | - | 2 | 332 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC299271 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181630.2 | NP_853661.1 | 2 | 332 | ||
GenBank mRNA | AY113703.1 | - | 2 | 332 |