siRNA Details
Gene Symbol: Mustn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181368.3 | NP_852033.1 | 2 | 365 | ||
GenBank mRNA | AY254906.2 | - | 2 | 365 | ||
BC158570.1 | - | 2 | 380 | 9092791 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Mustn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181368.3 | NP_852033.1 | 2 | 365 | ||
GenBank mRNA | AY254906.2 | - | 2 | 365 | ||
BC158570.1 | - | 2 | 380 | 9092791 |