siRNA Details
Gene Symbol: Rbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012640.2 | NP_036772.2 | 2 | 180 | ||
XM_006243607.1 | XP_006243669.1 | 3 | 360 | |||
GenBank mRNA | M13949.1 | - | 2 | 180 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Rbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012640.2 | NP_036772.2 | 2 | 180 | ||
XM_006243607.1 | XP_006243669.1 | 3 | 360 | |||
GenBank mRNA | M13949.1 | - | 2 | 180 |