siRNA Details
| Gene Symbol: Klrd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012745.2 | NP_036877.1 | 5 | 627 | ||
| XM_006237078.2 | XP_006237140.1 | 5 | 745 | |||
| GenBank mRNA | AF009133.1 | - | 5 | 384 | ||
| BC086393.1 | - | 5 | 627 | 7303354 | ||

Klrd1
Genomic Map