siRNA Details
| Gene Symbol: Ccnd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012766.1 | NP_036898.1 | 1 | 225 | ||
| GenBank mRNA | BC085763.1 | - | 1 | 258 | 7189593 | |
| BC089819.1 | - | 415 | 7384331 | |||
| D16309.1 | - | 1 | 225 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccnd3| Gene Symbol: Ccnd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012766.1 | NP_036898.1 | 1 | 225 | ||
| GenBank mRNA | BC085763.1 | - | 1 | 258 | 7189593 | |
| BC089819.1 | - | 415 | 7384331 | |||
| D16309.1 | - | 1 | 225 | |||