siRNA Details
| Gene Symbol: Tnp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017057.2 | NP_058753.2 | 1 | 130 | ||
| GenBank mRNA | BC078849.1 | - | 1 | 130 | 7131008 | |
| U52958.1 | - | 1 | 76 | |||
| X14776.1 | - | 1 | 118 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tnp2| Gene Symbol: Tnp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017057.2 | NP_058753.2 | 1 | 130 | ||
| GenBank mRNA | BC078849.1 | - | 1 | 130 | 7131008 | |
| U52958.1 | - | 1 | 76 | |||
| X14776.1 | - | 1 | 118 | |||