siRNA Details
| Gene Symbol: Pts | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017220.1 | NP_058916.1 | 2 | 150 | ||
| GenBank mRNA | BC059140.1 | - | 2 | 133 | 6888665 | |
| M77850.1 | - | 2 | 150 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pts| Gene Symbol: Pts | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017220.1 | NP_058916.1 | 2 | 150 | ||
| GenBank mRNA | BC059140.1 | - | 2 | 133 | 6888665 | |
| M77850.1 | - | 2 | 150 | |||